Trimethylsilanol
Trimethylsilanol
Trimethylsilanol
Thermo Scientific Chemicals

Trimethylsilanol

CAS: 1066-40-6 | C3H9LiOSi | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 458570250
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
140.00
Save 20.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
140.00
Save 20.35
Each
Chemical Identifiers
CAS1066-40-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only