4-Bromoisoxazole, 95%
4-Bromoisoxazole, 95%
4-Bromoisoxazole, 95%
Thermo Scientific Chemicals

4-Bromoisoxazole, 95%

CAS: 97925-43-4 | C3H2BrNO | 147.96 g/mol
Have Questions?
Catalog NumberQuantity
4587300101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 458730010
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.00
Save 5.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.00
Save 5.35
Each
Chemical Identifiers
CAS97925-43-4
IUPAC Name4-bromo-1,2-oxazole
Molecular FormulaC3H2BrNO
InChI KeyIRDVIGFQMLUJAM-UHFFFAOYSA-N
SMILESBrC1=CON=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94.0 %
Appearance (Color)White to yellow
Appearance (Form)Low melting solid
Infrared spectrumConforms
Melting point37°C to 43°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only