4-Azabenzimidazole formate, 97%, Thermo Scientific Chemicals
4-Azabenzimidazole formate, 97%, Thermo Scientific Chemicals
4-Azabenzimidazole formate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Azabenzimidazole formate, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
4588200101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 458820010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Azabenzimidazole formate
Elemental Analysis48.87 to max. 52.95% min. (Carbon) 24.42 to max. 26.46% min. (Nitrogen)
Formula Weight165.15
Infrared SpectrumAuthentic
Melting Point77.0°C to 78.0°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only