Doxycycline monohydrate, 97%
Doxycycline monohydrate, 97%
Doxycycline monohydrate, 97%
Thermo Scientific Chemicals

Doxycycline monohydrate, 97%

Have Questions?
Catalog NumberQuantity
4588900505g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 458890050
Price (USD)
103.00
Each
Quantity:
5g
Request bulk or custom format
Price (USD)
103.00
Each
Chemical Identifiers
CAS17086-28-1
IUPAC Name(2Z,4S,4aR,5S,5aR,6R,12aS)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-5,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione hydrate
Molecular FormulaC22H26N2O9
InChI KeyADAURKPGCFUREH-UQVCFKGQSA-N
SMILESO.C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H](N(C)C)C(=O)C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Water3.5 to 4.5 %
HPLC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only