Iodomethane, 99%, stabilized
Iodomethane, 99%, stabilized
Iodomethane, 99%, stabilized
Thermo Scientific Chemicals

Iodomethane, 99%, stabilized

CAS: 74-88-4 | CH3I | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 459015000
Price (USD)
228.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
228.00
Each
Chemical Identifiers
CAS74-88-4
IUPAC Nameiodomethane
Molecular FormulaCH3I
InChI KeyINQOMBQAUSQDDS-UHFFFAOYSA-N
SMILESCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
Infrared spectrumConforms
Refractive index1.5290 to 1.5310 (20°C, 589 nm)
GC>=98.5 %
StabilizerCopper
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only