Pyrrolidine-3-carboxylic acid, 98%
Pyrrolidine-3-carboxylic acid, 98%
Pyrrolidine-3-carboxylic acid, 98%
Thermo Scientific Chemicals

Pyrrolidine-3-carboxylic acid, 98%

CAS: 59378-87-9 | C5H9NO2 | 115.13 g/mol
Have Questions?
Catalog NumberQuantity
4591100505 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 459110050
Price (USD)
263.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
263.00
Each
Chemical Identifiers
CAS59378-87-9
IUPAC Namepyrrolidine-3-carboxylic acid
Molecular FormulaC5H9NO2
InChI KeyJAEIBKXSIXOLOL-UHFFFAOYNA-N
SMILESOC(=O)C1CCNC1
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis>=11.86 to =<12.47 % Nitrogen
Appearance (Color)White to light orange
Appearance (Form)Powder or solid
Infrared spectrumConforms
Elemental Analysis>=50.86 to =<53.47 % Carbon
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only