4,4',4″-Tris(carbazol-9-yl)triphenylamine, 99%
4,4',4″-Tris(carbazol-9-yl)triphenylamine, 99%
4,4',4″-Tris(carbazol-9-yl)triphenylamine, 99%
Thermo Scientific Chemicals

4,4',4″-Tris(carbazol-9-yl)triphenylamine, 99%

CAS: 139092-78-7 | C54H36N4 | 740.91 g/mol
Have Questions?
Catalog NumberQuantity
4591800101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 459180010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS139092-78-7
IUPAC Name4-(9H-carbazol-9-yl)-N,N-bis[4-(9H-carbazol-9-yl)phenyl]aniline
Molecular FormulaC54H36N4
InChI KeyAWXGSYPUMWKTBR-UHFFFAOYSA-N
SMILESC1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=C(C=C1)N(C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=98.5 %
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only