Sodium ferrocyanide decahydrate
Sodium ferrocyanide decahydrate
Sodium ferrocyanide decahydrate
Thermo Scientific Chemicals

Sodium ferrocyanide decahydrate

CAS: 14434-22-1 ∣ C6H20FeN6Na4O10 ∣ 484.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 459290010
Price (USD)
60.40
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
60.40
Each
Chemical Identifiers
CAS14434-22-1
Molecular FormulaC6H20FeN6Na4O10
InChI KeyJFSUDVTVQZUDOP-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Molecular Weight (g/mol)484.06
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder or crystals
Chloride (Cl)=<0.1 %
Titration>=98.5 %
Loss on drying=<1 % (free moisture)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only