Risperidone, 99%
Risperidone, 99%
Risperidone, 99%
Thermo Scientific Chemicals

Risperidone, 99%

CAS: 106266-06-2 | C23H27FN4O2 | 410.49 g/mol
Have Questions?
Catalog NumberQuantity
4593900101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 459390010
Price (USD)
68.65
Special offer
Online exclusive
Ends: 28-Dec-2026
80.00
Save 11.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
68.65
Special offer
Online exclusive
Ends: 28-Dec-2026
80.00
Save 11.35
Each
Chemical Identifiers
CAS106266-06-2
IUPAC Name3-{2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl}-2-methyl-4H,6H,7H,8H,9H-pyrido[1,2-a]pyrimidin-4-one
Molecular FormulaC23H27FN4O2
InChI KeyRAPZEAPATHNIPO-UHFFFAOYSA-N
SMILESCC1=C(CCN2CCC(CC2)C2=NOC3=CC(F)=CC=C23)C(=O)N2CCCCC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point170°C to 174°C
HPLC>=98.5 %
Loss on drying=<0.5 %
Appearance (Color)White to light yellow
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only