N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, 98%
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, 98%
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, 98%
Thermo Scientific Chemicals

N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, 98%

CAS: 15163-36-7 | C15H33NO3S | 307.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 459680050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS15163-36-7
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Solubility(10% in water) Clear colorless solution
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only