4,6-Diaminopyrimidine, 98%, Thermo Scientific Chemicals
4,6-Diaminopyrimidine, 98%, Thermo Scientific Chemicals
4,6-Diaminopyrimidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,6-Diaminopyrimidine, 98%, Thermo Scientific Chemicals

CAS: 2434-56-2 | C4H6N4 | 110.1 g/mol
Have Questions?
Catalog NumberQuantity
4598000505 g
Catalog number 459800050
Price (USD)
142.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
142.00
Each
Chemical Identifiers
CAS2434-56-2
IUPAC Namepyrimidine-4,6-diamine
Molecular FormulaC4H6N4
InChI KeyMISVBCMQSJUHMH-UHFFFAOYSA-N
SMILESNC1=CC(N)=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Water=<1 %
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only