1H-Indole-2-methanol, 96%
1H-Indole-2-methanol, 96%
1H-Indole-2-methanol, 96%
Thermo Scientific Chemicals

1H-Indole-2-methanol, 96%

CAS: 24621-70-3 | C9H9NO | 147.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 459810050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS24621-70-3
IUPAC Name(1H-indol-2-yl)methanol
Molecular FormulaC9H9NO
InChI KeyXEEANGGQJOWRTG-UHFFFAOYSA-N
SMILESOCC1=CC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light yellow to tan
Appearance (Form)Solid
Melting point72°C to 78°C
GC>=95.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only