CD1d Monoclonal Antibody (1B1), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD1d Monoclonal Antibody (1B1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD1d Monoclonal Antibody (1B1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0011-82, 46-0011-80)