CD1c Monoclonal Antibody (L161), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD1c Monoclonal Antibody (L161), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD1c Monoclonal Antibody (L161), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0015-42, 46-0015-41)