CD2 Monoclonal Antibody (RPA-2.10), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD2 Monoclonal Antibody (RPA-2.10), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD2 Monoclonal Antibody (RPA-2.10), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0029-41, 46-0029-42)