CD3 Monoclonal Antibody (17A2), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD3 Monoclonal Antibody (17A2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (17A2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0032-80, 46-0032-82)