CD3 Monoclonal Antibody (SK7), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD3 Monoclonal Antibody (SK7), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (SK7), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0036-41, 46-0036-42)