CD3 Monoclonal Antibody (OKT3), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD3 Monoclonal Antibody (OKT3), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (OKT3), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0037-41, 46-0037-42)