CD4 Monoclonal Antibody (OX35), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD4 Monoclonal Antibody (OX35), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (OX35), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0040-82)