CD4 Monoclonal Antibody (RM4-5), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD4 Monoclonal Antibody (RM4-5), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (RM4-5), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0042-82, 46-0042-80)