CD5 Monoclonal Antibody (UCHT2), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD5 Monoclonal Antibody (UCHT2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD5 Monoclonal Antibody (UCHT2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0059-42)