CD7 Monoclonal Antibody (4H9), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD7 Monoclonal Antibody (4H9), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD7 Monoclonal Antibody (4H9), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0078-42)