CD8a Monoclonal Antibody (53-6.7), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD8a Monoclonal Antibody (53-6.7), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (53-6.7), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0081-82, 46-0081-80)