CD8a Monoclonal Antibody (SK1), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD8a Monoclonal Antibody (SK1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (SK1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0087-42, 46-0087-41)