CD11b (activation epitope) Monoclonal Antibody (CBRM1/5), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD11b (activation epitope) Monoclonal Antibody (CBRM1/5), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD11b (activation epitope) Monoclonal Antibody (CBRM1/5), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0113-41, 46-0113-42)