CD11c Monoclonal Antibody (3.9), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD11c Monoclonal Antibody (3.9), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD11c Monoclonal Antibody (3.9), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0116-41)