CD11b Monoclonal Antibody (ICRF44), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD11b Monoclonal Antibody (ICRF44), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD11b Monoclonal Antibody (ICRF44), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0118-41, 46-0118-42)