CD14 Monoclonal Antibody (Sa2-8), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD14 Monoclonal Antibody (Sa2-8), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0141-82, 46-0141-80)