CD14 Monoclonal Antibody (61D3), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD14 Monoclonal Antibody (61D3), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD14 Monoclonal Antibody (61D3), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0149-42, 46-0149-41)