CD15 Monoclonal Antibody (MMA), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD15 Monoclonal Antibody (MMA), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD15 Monoclonal Antibody (MMA), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0158-42, 46-0158-41)