CD15 Monoclonal Antibody (HI98), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD15 Monoclonal Antibody (HI98), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD15 Monoclonal Antibody (HI98), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0159-42, 46-0159-41)