CD16/CD32 Monoclonal Antibody (93), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD16/CD32 Monoclonal Antibody (93), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD16/CD32 Monoclonal Antibody (93), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0161-82, 46-0161-80)