CD16 Monoclonal Antibody (3G8), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD16 Monoclonal Antibody (3G8), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0166-41, 46-0166-42)