CD19 Monoclonal Antibody (SJ25C1), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD19 Monoclonal Antibody (SJ25C1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (SJ25C1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0198-42)