CD20 Monoclonal Antibody (QCH6A7), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD20 Monoclonal Antibody (QCH6A7), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD20 Monoclonal Antibody (QCH6A7), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0203-80, 46-0203-82)