CD21/CD35 Monoclonal Antibody (eBio4E3 (4E3)), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD21/CD35 Monoclonal Antibody (eBio4E3 (4E3)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD21/CD35 Monoclonal Antibody (eBio4E3 (4E3)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0212-82)