CD21 Monoclonal Antibody (HB5), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD21 Monoclonal Antibody (HB5), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD21 Monoclonal Antibody (HB5), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0219-41, 46-0219-42)