CD22 Monoclonal Antibody (eBio4KB128 (4KB128)), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD22 Monoclonal Antibody (eBio4KB128 (4KB128)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD22 Monoclonal Antibody (eBio4KB128 (4KB128)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0229-42)