CD23 Monoclonal Antibody (B3B4), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD23 Monoclonal Antibody (B3B4), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD23 Monoclonal Antibody (B3B4), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0232-80)