CD24 Monoclonal Antibody (M1/69), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD24 Monoclonal Antibody (M1/69), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD24 Monoclonal Antibody (M1/69), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0242-80, 46-0242-82)