CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0247-42, 46-0247-41)