CD25 Monoclonal Antibody (CD25-4E3), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD25 Monoclonal Antibody (CD25-4E3), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (CD25-4E3), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0257-41, 46-0257-42)