CD27 Monoclonal Antibody (LG.7F9), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD27 Monoclonal Antibody (LG.7F9), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD27 Monoclonal Antibody (LG.7F9), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0271-80, 46-0271-82)