CD27 Monoclonal Antibody (O323), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD27 Monoclonal Antibody (O323), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD27 Monoclonal Antibody (O323), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0279-42, 46-0279-41)