CD29 (Integrin beta 1) Monoclonal Antibody (eBioHMb1-1 (HMb1-1)), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD29 (Integrin beta 1) Monoclonal Antibody (eBioHMb1-1 (HMb1-1)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD29 (Integrin beta 1) Monoclonal Antibody (eBioHMb1-1 (HMb1-1)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0291-82, 46-0291-80)