CD31 (PECAM-1) Monoclonal Antibody (WM-59 (WM59)), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD31 (PECAM-1) Monoclonal Antibody (WM-59 (WM59)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD31 (PECAM-1) Monoclonal Antibody (WM-59 (WM59)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0319-42)