SIGLEC H Monoclonal Antibody (eBio440c), PerCP-eFluor™ 710, eBioscience™ - FAQs
SIGLEC H Monoclonal Antibody (eBio440c), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for SIGLEC H Monoclonal Antibody (eBio440c), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0333-82, 46-0333-80)