CD34 Monoclonal Antibody (4H11), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD34 Monoclonal Antibody (4H11), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD34 Monoclonal Antibody (4H11), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0349-42, 46-0349-41)