CD38 Monoclonal Antibody (90), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD38 Monoclonal Antibody (90), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD38 Monoclonal Antibody (90), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0381-80, 46-0381-82)