CD38 Monoclonal Antibody (HB7), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD38 Monoclonal Antibody (HB7), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD38 Monoclonal Antibody (HB7), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0388-41, 46-0388-42)